Gene Transcripts

Gene Symbol
DIRAS2
Entrez Gene ID
54769
Gene Name
DIRAS family GTPase 2
Gene Aliases
Di-Ras2
Chromosome Location
Chr. 9: 90609832 - 90643105 on Build GRCh38
UniGene ID
Hs.165636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017594.4 NP_060064.2 2 1770
GenBank mRNA AB076889.1 - 1 1410
AK122986.1 - 2 1491
AK124628.1 - 3 1435
AL137308.1 - 1 410
BC008065.2 - 2 1450 3029902

View Full Product Details