Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_182919.3 | NP_891549.1 | 2 | 2054 | |
GenBank mRNA | AB086380.1 | - | 1 | 1848 | |
AB093555.1 | - | 1 | 1786 | ||
AB097023.1 | - | 1 | 1786 | ||
AB446484.1 | - | 1 | 1786 | ||
AF070530.1 | - | 1 | 900 | ||
AF492646.1 | - | 1 | 1374 | ||
BC009860.2 | - | 1 | 720 | ||
BC136556.1 | - | 1 | 1840 | 9020546 | |
BC136557.1 | - | 1 | 1840 | 9020547 |