Gene Transcripts

Gene Symbol
UFSP1
Entrez Gene ID
402682
Gene Name
UFM1 specific peptidase 1 (inactive)
Gene Aliases
UFSP
Chromosome Location
Chr. 7: 100888723 - 100889718 on Build GRCh38
UniGene ID
Hs.534845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015072.3 NP_001015072.2 1 869
GenBank mRNA AK292467.1 - 1 869
BC017456.2 - 1 804
BC067904.1 - 1 832 5310615

View Full Product Details