Gene Transcripts

Gene Symbol
Mfsd2a
Entrez Gene ID
76574
Gene Name
major facilitator superfamily domain containing 2A
Gene Aliases
1700018O18Rik, Mfsd2, NLS1
Chromosome Location
Chr. 4: 122946850 - 122961195 on Build GRCm38
UniGene ID
Mm.331842
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029662.2 NP_083938.2 6, 7 888
GenBank mRNA AK006096.1 - 6, 7 887
BC046793.1 - 6, 7 870 5717701
BC060526.1 - 6, 7 859 5707105

View Full Product Details