Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099666.1 | NP_001093136.1 | 8 | 1244 | |
XM_005251976.4 | XP_005252033.1 | 7 | 1415 | ||
XM_005251977.4 | XP_005252034.1 | 8 | 1305 | ||
XM_011518639.1 | XP_011516941.1 | 7 | 1289 | ||
XM_011518640.1 | XP_011516942.1 | 8 | 1465 | ||
GenBank mRNA | AK127511.1 | - | 2 | 579 | |
AL832683.1 | - | 5 | 1069 | ||
BC053622.1 | - | 8 | 1226 |