Gene Transcripts

Gene Symbol
PCDHB13
Entrez Gene ID
56123
Gene Name
protocadherin beta 13
Gene Aliases
PCDH-BETA13
Chromosome Location
Chr. 5: 141213919 - 141217421 on Build GRCh38
UniGene ID
Hs.283803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018933.3 NP_061756.1 1 3205
GenBank mRNA AF217745.1 - 1 3000

View Full Product Details