Gene Transcripts
- Gene Symbol
- SNX18
- Entrez Gene ID
- 112574
- Gene Name
- sorting nexin 18
- Gene Aliases
- SH3PX2, SH3PXD3B, SNAG1
- Chromosome Location
- Chr. 5: 54517759 - 54619249 on Build GRCh38
- UniGene ID
- Hs.432755
- Species
- Homo sapiens
- Gene Symbol
- LINC00661
- Entrez Gene ID
- 126536
- Gene Name
- long intergenic non-protein coding RNA 661
- Gene Aliases
- Chromosome Location
- Chr. 19: 16015634 - 16027462 on Build GRCh38
- UniGene ID
- Hs.131824
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026828.1 |
|
9 |
3176 |
|
GenBank mRNA |
AK128240.1 |
- |
4 |
2695 |
|
CR749400.1 |
- |
9 |
3187 |
|
View Full Product Details