Gene Transcripts

Gene Symbol
Gtpbp10
Entrez Gene ID
207704
Gene Name
GTP-binding protein 10 (putative)
Gene Aliases
4930545J22Rik, BC034507
Chromosome Location
Chr. 5: 5537450 - 5559774 on Build GRCm38
UniGene ID
Mm.323984
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153116.1 NP_694756.1 10 1684
GenBank mRNA AK139595.1 - 10 1698
BC034507.1 - 10 1710 4222393

View Full Product Details