Gene Transcripts

Gene Symbol
Acmsd
Entrez Gene ID
266645
Gene Name
amino carboxymuconate semialdehyde decarboxylase
Gene Aliases
Chromosome Location
Chr. 1: 127729413 - 127767564 on Build GRCm38
UniGene ID
Mm.25735
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033041.2 NP_001028213.1 3 213
GenBank mRNA AB071419.1 - 3 140
AK143962.1 - 3 215
BC139281.1 - 3 164 8862303

View Full Product Details