Gene Transcripts

Gene Symbol
WDR25
Entrez Gene ID
79446
Gene Name
WD repeat domain 25
Gene Aliases
C14orf67
Chromosome Location
Chr. 14: 100376410 - 100530303 on Build GRCh38
UniGene ID
Hs.497600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161476.1 NP_001154948.1 4, 5 1202
NM_024515.4 NP_078791.3 4, 5 1333
XM_005268056.4 XP_005268113.1 4, 5 1209
XM_005268057.3 XP_005268114.1 3, 4 359
XM_005268058.4 XP_005268115.1 3, 4 373
XM_011537150.2 XP_011535452.1 4, 5 1456
XM_011537151.2 XP_011535453.2 3, 4 812
XM_017021639.1 XP_016877128.1 3, 4 361
GenBank mRNA AK291960.1 - 4, 5 1165
AY295082.1 - 4, 5 1216
BC003641.2 - 3, 4 439 3529568
BC007953.2 - 2, 3 986
CT003969.1 - 3, 4 365

View Full Product Details