Gene Transcripts

Gene Symbol
PRR7
Entrez Gene ID
80758
Gene Name
proline rich 7 (synaptic)
Gene Aliases
Chromosome Location
Chr. 5: 177445936 - 177456701 on Build GRCh38
UniGene ID
Hs.534492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174101.1 NP_001167572.1 3 1222
NM_001174102.1 NP_001167573.1 3 1139
NM_030567.4 NP_085044.2 4 1307
XM_011534662.1 XP_011532964.1 3 7045
XM_011534663.2 XP_011532965.1 3 1783
XM_017009896.1 XP_016865385.1 2 7833
XM_017009897.1 XP_016865386.1 3 2015
GenBank mRNA BC004261.1 - 4 1139
BC021240.1 - 3 1130 3603954
BC024233.2 - 4 1311 4697841
BC033649.1 - 2 897 5538503

View Full Product Details