Gene Transcripts

Gene Symbol
PRR7
Entrez Gene ID
80758
Gene Name
proline rich 7 (synaptic)
Gene Aliases
Chromosome Location
Chr. 5: 177445936 - 177456701 on Build GRCh38
UniGene ID
Hs.534492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174101.1 NP_001167572.1 3 1335
NM_001174102.1 NP_001167573.1 3 1252
NM_030567.4 NP_085044.2 4 1420
XM_011534662.1 XP_011532964.1 3 7158
XM_011534663.2 XP_011532965.1 3 1896
XM_017009896.1 XP_016865385.1 2 7946
XM_017009897.1 XP_016865386.1 3 2128
GenBank mRNA BC004261.1 - 4 1252
BC021240.1 - 3 1243 3603954
BC024233.2 - 4 1424 4697841

View Full Product Details