Gene Transcripts

Gene Symbol
GTPBP10
Entrez Gene ID
85865
Gene Name
GTP binding protein 10
Gene Aliases
ObgH2, UG0751c10
Chromosome Location
Chr. 7: 90346665 - 90391455 on Build GRCh38
UniGene ID
Hs.593547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042717.2 NP_001036182.1 8 1227
NM_033107.3 NP_149098.2 10 1464
GenBank mRNA AF351613.1 - 8 1228
AK095561.1 - 10 1454
AK125553.1 - 1 461
AK294325.1 - 8 1177
BC144162.1 - 8 1173
CR933597.1 - 9 1278

View Full Product Details