Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052855.3 | NP_443087.1 | 2 | 520 | |
XM_017025296.1 | XP_016880785.1 | 2 | 267 | ||
GenBank mRNA | AK314010.1 | - | 2 | 267 | |
BC012978.2 | - | 2 | 512 | 3834331 | |
CR627413.1 | - | 2 | 292 | ||
EU164539.1 | - | 2 | 512 |