Gene Transcripts

Gene Symbol
SYTL5
Entrez Gene ID
94122
Gene Name
synaptotagmin like 5
Gene Aliases
slp5
Chromosome Location
Chr. X: 37888915 - 38128820 on Build GRCh38
UniGene ID
Hs.662334
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163334.1 NP_001156806.1 1, 2 483
NM_001163335.1 NP_001156807.1 3, 4 661
NM_138780.2 NP_620135.1 2, 3 570
XM_011544001.2 XP_011542303.1 4, 5 922
XM_011544002.2 XP_011542304.1 2, 3 556
XM_017029972.1 XP_016885461.1 5, 6 1088
XM_017029973.1 XP_016885462.1 4, 5 1155
XM_017029974.1 XP_016885463.1 3, 4 783
GenBank mRNA AB080222.1 - 1, 2 127
AK125106.1 - 2, 3 539
BC131585.1 - 1, 2 127 40113205
BX647688.1 - 3, 4 663

View Full Product Details