Gene Transcripts

Gene Symbol
THAP6
Entrez Gene ID
152815
Gene Name
THAP domain containing 6
Gene Aliases
Chromosome Location
Chr. 4: 75513951 - 75547334 on Build GRCh38
UniGene ID
Hs.479971
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317791.1 NP_001304720.1 2, 3 180
NM_144721.5 NP_653322.1 2, 3 180
XM_005262774.3 XP_005262831.1 2, 3 182
XM_006714109.3 XP_006714172.1 2, 3 182
XM_011531666.2 XP_011529968.1 2, 3 617
XM_011531667.2 XP_011529969.1 2, 3 140
XM_017007800.1 XP_016863289.1 2, 3 346
XM_017007801.1 XP_016863290.1 2, 3 182
GenBank mRNA AK303658.1 - 2, 3 140
AK313765.1 - 2, 3 564
AL833350.1 - 2, 3 140
BC022989.2 - 1, 2 94 5113206
BX647579.1 - 2, 3 140
DC308535.1 - 2, 3 181

View Full Product Details