Gene Transcripts

Gene Symbol
NLRP11
Entrez Gene ID
204801
Gene Name
NLR family pyrin domain containing 11
Gene Aliases
CLR19.6, NALP11, NOD17, PAN10, PYPAF6, PYPAF7
Chromosome Location
Chr. 19: 55785397 - 55836762 on Build GRCh38
UniGene ID
Hs.375039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145007.3 NP_659444.2 4 953
GenBank mRNA AK310394.1 - 4 630
AY095145.1 - 2 341
AY154466.1 - 1 242
AY168967.1 - 2 330
AY226382.1 - 1 242
BC034730.1 - 2 330 4341197

View Full Product Details