Gene Transcripts

Gene Symbol
C7orf33
Entrez Gene ID
202865
Gene Name
chromosome 7 open reading frame 33
Gene Aliases
Chromosome Location
Chr. 7: 148590565 - 148615860 on Build GRCh38
UniGene ID
Hs.352212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145304.3 NP_660347.1 1 535
GenBank mRNA BC021251.2 - 1 535

View Full Product Details