Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005996.3 | NP_005987.3 | 1 | 1296 | |
NM_016569.3 | NP_057653.3 | 1 | 1296 | ||
GenBank mRNA | AF002228.1 | - | 1 | 332 | |
AF170708.2 | - | 1 | 592 | ||
AF216750.1 | - | 1 | 448 | ||
AK054604.1 | - | 1 | 1299 | ||
AK304639.1 | - | 1 | 566 | ||
BC025258.1 | - | 1 | 722 | 5458526 |