Gene Transcripts

Gene Symbol
FBXO39
Entrez Gene ID
162517
Gene Name
F-box protein 39
Gene Aliases
CT144, Fbx39
Chromosome Location
Chr. 17: 6776184 - 6798929 on Build GRCh38
UniGene ID
Hs.368364
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153230.2 NP_694962.1 3, 4 1328
XM_011523697.1 XP_011521999.1 3, 4 1404
GenBank mRNA BC034782.1 - 3, 4 1308
DB524242.1 - 1, 2 144

View Full Product Details