Gene Transcripts

Gene Symbol
CNFN
Entrez Gene ID
84518
Gene Name
cornifelin
Gene Aliases
PLAC8L2
Chromosome Location
Chr. 19: 42387019 - 42390317 on Build GRCh38
UniGene ID
Hs.148590
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032488.3 NP_115877.2 4 390
XM_005259332.2 XP_005259389.1 5 495
XM_011527396.1 XP_011525698.1 5 490
XM_011527397.1 XP_011525699.1 5 436
GenBank mRNA AB049591.2 - 4 385
BC101197.1 - 5 446 40021735
BC101198.2 - 4 366 40021738

View Full Product Details