Gene Transcripts
- Gene Symbol
- Klk1c2
- Entrez Gene ID
- 24841
- Gene Name
- kallikrein 1-related peptidase C2
- Gene Aliases
- KLK2, Ton, rGK-2
- Chromosome Location
- Chr. 1: 99981045 - 99985422 on Build Rnor_6.0
- UniGene ID
- Rn.9882
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012677.1 |
NP_036809.1
|
4 |
650 |
|
GenBank mRNA |
M11565.1 |
- |
4 |
650 |
|
View Full Product Details