Gene Transcripts

Gene Symbol
Ssr4
Entrez Gene ID
29435
Gene Name
signal sequence receptor, delta
Gene Aliases
Chromosome Location
Chr. X: 156995763 - 156999709 on Build Rnor_6.0
UniGene ID
Rn.1999
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017199.1 NP_058895.1 2, 3 345
XM_006229571.2 XP_006229633.1 2, 3 401
GenBank mRNA BC058482.1 - 2, 3 258
Z19087.1 - 2, 3 345

View Full Product Details