Gene Transcripts

Gene Symbol
Omt2a
Entrez Gene ID
18379
Gene Name
oocyte maturation, alpha
Gene Aliases
OM2a
Chromosome Location
Chr. 9: 78311972 - 78314797 on Build GRCm38
UniGene ID
Mm.335311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111286.2 NP_001104756.1 1 238
GenBank mRNA AK139546.1 - 1 206
DV649105.1 - 1 238

View Full Product Details