Gene Transcripts

Gene Symbol
Krtap9-5
Entrez Gene ID
435286
Gene Name
keratin associated protein 9-5
Gene Aliases
Chromosome Location
Chr. 11: 99948475 - 99949551 on Build GRCm38
UniGene ID
Mm.457964
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085527.1 NP_001078996.1 1 1067

View Full Product Details