Gene Transcripts

Gene Symbol
Rhox3f
Entrez Gene ID
621852
Gene Name
reproductive homeobox 3F
Gene Aliases
OTTMUSG00000017155, Rhox3.6, Rhox3e
Chromosome Location
Chr. X: 37581352 - 37585496 on Build GRCm38
UniGene ID
Mm.490237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040089.3 NP_001035178.3 4 627
GenBank mRNA AY147207.1 - 520
BC145759.1 - 497

View Full Product Details