Gene Transcripts
- Gene Symbol
- Prl7b1
- Entrez Gene ID
- 266804
- Gene Name
- prolactin family 7, subfamily b, member 1
- Gene Aliases
- Prlpn
- Chromosome Location
- Chr. 17: 39153431 - 39161644 on Build Rnor_6.0
- UniGene ID
- Rn.79989
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153738.1 |
NP_714960.1
|
4, 5 |
395 |
|
GenBank mRNA |
AF525159.1 |
- |
4, 5 |
395 |
|
View Full Product Details