Gene Transcripts

Gene Symbol
Cyp4b1
Entrez Gene ID
13120
Gene Name
cytochrome P450, family 4, subfamily b, polypeptide 1
Gene Aliases
Chromosome Location
Chr. 4: 115624728 - 115647705 on Build GRCm38
UniGene ID
Mm.1840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007823.2 NP_031849.1 2 233
GenBank mRNA AK143890.1 - 2 246
AK165445.1 - 2 249
BC008996.1 - 2 237 3967680
D50834.1 - 2 233

View Full Product Details