Gene Transcripts

Gene Symbol
Ptdss1
Entrez Gene ID
19210
Gene Name
phosphatidylserine synthase 1
Gene Aliases
AU044268, AW539008, PSS-1, mKIAA0024
Chromosome Location
Chr. 13: 66932830 - 66998401 on Build GRCm38
UniGene ID
Mm.281464
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008959.3 NP_032985.2 1 776
GenBank mRNA AF042731.1 - 1 191
AK036990.1 - 1 238
AK054101.1 - 1 215
AK085857.1 - 1 777
AK086136.1 - 1 391
AK088048.1 - 1 258
AK137681.1 - 1 446
AK172873.1 - 1 753
BC003260.1 - 1 413 3501141

View Full Product Details