Gene Transcripts
- Gene Symbol
- Nid1
- Entrez Gene ID
- 18073
- Gene Name
- nidogen 1
- Gene Aliases
- A630025O17, Nid, entactin, entactin-1, nidogen-1
- Chromosome Location
- Chr. 13: 13437602 - 13512275 on Build GRCm38
- UniGene ID
- Mm.4691
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010917.2 |
NP_035047.2
|
2, 3 |
624 |
|
GenBank mRNA |
AK041633.1 |
- |
2, 3 |
560 |
|
AK049585.1 |
- |
2, 3 |
560 |
|
AK084876.1 |
- |
2, 3 |
563 |
|
AK144878.1 |
- |
2, 3 |
589 |
|
AK166779.1 |
- |
2, 3 |
560 |
|
BC052939.1 |
- |
2, 3 |
619 |
|
BC131668.1 |
- |
2, 3 |
535 |
|
BC131669.1 |
- |
2, 3 |
535 |
40110859 |
X14194.1 |
- |
2, 3 |
535 |
|
X14480.1 |
- |
2, 3 |
545 |
|
View Full Product Details