Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159706.1 | NP_001153178.1 | 2 | 296 | |
NM_016770.3 | NP_058050.3 | 2 | 296 | ||
GenBank mRNA | AF026380.1 | - | 2 | 278 | |
AK002920.1 | - | 2 | 263 | ||
AK090289.1 | - | 297 | |||
BB872570.1 | - | 2 | 297 | ||
BC119604.1 | - | 2 | 240 | 40060385 | |
BC119605.1 | - | 2 | 240 | 40060386 |