Gene Transcripts

Gene Symbol
Psmd12
Entrez Gene ID
66997
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 12
Gene Aliases
1500002F15Rik, AI480719, P55
Chromosome Location
Chr. 11: 107479528 - 107498036 on Build GRCm38
UniGene ID
Mm.491171
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025894.2 NP_080170.1 11 1404
GenBank mRNA AK005114.1 - 11 1438
AK007619.1 - 11 1449
AK007680.1 - 11 1378
AK008646.1 - 11 1403
AK009969.1 - 11 1407
AK017843.1 - 11 1421
AK150910.1 - 11 1427
AK151242.1 - 11 1405
AK152041.1 - 11 1427
AK152240.1 - 11 1436
AK160283.1 - 11 1427
AK167236.1 - 11 1411
BC004694.1 - 11 1422 3498952

View Full Product Details