Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028715.3 | NP_082991.3 | 10 | 826 | |
GenBank mRNA | AK014382.1 | - | 10 | 811 | |
AK088957.1 | - | 10 | 810 | ||
AK137333.1 | - | 10 | 813 | ||
AK172927.1 | - | 7 | 697 | ||
BC032207.1 | - | 9 | 802 | 5323527 |