Gene Transcripts

Gene Symbol
Lin37
Entrez Gene ID
75660
Gene Name
lin-37 homolog (C. elegans)
Gene Aliases
1810054G18Rik
Chromosome Location
Chr. 7: 30555441 - 30559719 on Build GRCm38
UniGene ID
Mm.379242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290569.1 NP_001277498.1 6, 7 749
NM_029377.2 NP_083653.1 6, 7 623
GenBank mRNA AK007858.1 - 6, 7 624
AY428767.1 - 6, 7 473
BC013546.1 - 6, 7 605 4210417

View Full Product Details