Gene Transcripts

Gene Symbol
Msl3l2
Entrez Gene ID
73390
Gene Name
male-specific lethal 3-like 2 (Drosophila)
Gene Aliases
1700060H10Rik
Chromosome Location
Chr. 10: 56083827 - 56116880 on Build GRCm38
UniGene ID
Mm.181475
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163833.1 NP_001157305.1 2 1743
GenBank mRNA AK006842.1 - 1 1144
AK044179.1 - 1 1143
AK155382.1 - 2 1729
AK172141.1 - 125
BC013191.1 - 1 1345
BC037487.1 - 2 1754

View Full Product Details