Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166556.1 | NP_001160028.1 | 3 | 410 | |
NM_001166557.1 | NP_001160029.1 | 3 | 410 | ||
NM_147218.2 | NP_671751.2 | 3 | 410 | ||
GenBank mRNA | AF498361.1 | - | 3 | 362 | |
AK018242.1 | - | 3 | 395 | ||
AK040652.1 | - | 3 | 410 | ||
AK042934.1 | - | 3 | 382 | ||
AK053384.1 | - | 3 | 337 | ||
BC132417.1 | - | 3 | 247 | 40130694 | |
BC132419.1 | - | 3 | 247 | 40130696 |