Gene Transcripts

Gene Symbol
FABP5P3
Entrez Gene ID
220832
Gene Name
fatty acid binding protein 5 pseudogene 3
Gene Aliases
FABP5L3
Chromosome Location
Chr. 7: 152436895 - 152443016 on Build GRCh38
UniGene ID
Hs.647108
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002935.1 2 699
GenBank mRNA BX338084.2 - 2 699

Gene Symbol
FABP5
Entrez Gene ID
2171
Gene Name
fatty acid binding protein 5
Gene Aliases
E-FABP, EFABP, KFABP, PA-FABP, PAFABP
Chromosome Location
Chr. 8: 81280483 - 81284777 on Build GRCh38
UniGene ID
Hs.408061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001444.2 NP_001435.1 3 430
GenBank mRNA AK226036.1 - 3 379
AK311856.1 - 3 377
BC019385.2 - 3 363 3862100
BC070303.1 - 1 377 6496304
BT007449.1 - 1 317
M94856.1 - 3 365

View Full Product Details