Gene Transcripts

Gene Symbol
AQP4
Entrez Gene ID
361
Gene Name
aquaporin 4
Gene Aliases
MIWC, WCH4
Chromosome Location
Chr. 18: 26852038 - 26865844 on Build GRCh38
UniGene ID
Hs.315369
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317384.1 NP_001304313.1 2 413
NM_001317387.1 NP_001304316.1 2 413
NM_001650.5 NP_001641.1 2 413
NM_004028.4 NP_004019.1 1 540
XM_011525942.2 XP_011524244.1 2 455
GenBank mRNA AB209156.1 - 2 279
AK222684.1 - 2 325
BC022286.1 - 2 278 4717755
BI596912.1 - 2 313
BI667387.1 - 2 331
D63412.1 - 2 278
HQ901095.1 - 2 387
KF055862.1 - 2 258
U34845.1 - 1 476
U63622.1 - 2 297
U63623.1 - 1 201

View Full Product Details