Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001171092.1 | NP_001164563.1 | 2 | 977 | |
NM_001171095.1 | NP_001164566.1 | 2 | 746 | ||
NM_020384.3 | NP_065117.1 | 2 | 812 | ||
GenBank mRNA | AF177340.1 | - | 2 | 977 | |
AF250558.1 | - | 1 | 458 | ||
AK075371.1 | - | 2 | 688 | ||
AK075405.1 | - | 2 | 784 | ||
AK312515.1 | - | 2 | 746 | ||
AY358474.1 | - | 1 | 579 | ||
BC014424.1 | - | 1 | 579 | 4645075 | |
BC071747.1 | - | 2 | 785 | 30322852 |