Gene Transcripts

Gene Symbol
FLRT1
Entrez Gene ID
23769
Gene Name
fibronectin leucine rich transmembrane protein 1
Gene Aliases
SPG68
Chromosome Location
Chr. 11: 64035970 - 64119183 on Build GRCh38
UniGene ID
Hs.584876
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013280.4 NP_037412.2 2 1886
XM_005273861.1 XP_005273918.1 3 2772
XM_011544883.1 XP_011543185.1 2 1726
GenBank mRNA AF007139.1 - 1 261
AF169675.1 - 1 1545
AK289931.1 - 2 2203
BC018370.1 - 2 1886 4214494

View Full Product Details