Gene Transcripts

Gene Symbol
C3orf18
Entrez Gene ID
51161
Gene Name
chromosome 3 open reading frame 18
Gene Aliases
G20
Chromosome Location
Chr. 3: 50558025 - 50574835 on Build GRCh38
UniGene ID
Hs.517860
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171740.2 NP_001165211.1 2 623
NM_001171741.2 NP_001165212.1 2 418
NM_001171743.2 NP_001165214.2 2 462
NM_016210.4 NP_057294.2 3 712
XM_011533782.2 XP_011532084.1 3 688
XM_011533783.2 XP_011532085.1 2 578
XM_011533784.2 XP_011532086.1 3 466
XM_011533785.2 XP_011532087.1 2 404
XM_011533790.1 XP_011532092.1 3 688
XM_017006545.1 XP_016862034.1 3 688
XM_017006547.1 XP_016862036.1 2 623
GenBank mRNA AF188706.1 - 2 383
AK124438.1 - 2 463
AK308074.1 - 2 346
AK313128.1 - 3 586
AL833938.1 - 3 671
BC034766.1 - 2 496 5177784

View Full Product Details