Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001244676.1 | NP_001231605.1 | 3 | 465 | |
NM_018271.4 | NP_060741.3 | 2 | 303 | ||
XM_005264400.4 | XP_005264457.1 | 3 | 415 | ||
XM_005264401.4 | XP_005264458.1 | 3 | 384 | ||
XM_005264402.3 | XP_005264459.1 | 2 | 2204 | ||
XM_005264403.4 | XP_005264460.1 | 2 | 2204 | ||
XM_005264405.4 | XP_005264462.1 | 2 | 2204 | ||
XM_006712041.3 | XP_006712104.1 | 2 | 2204 | ||
XM_006712042.3 | XP_006712105.1 | 2 | 2204 | ||
XM_006712043.2 | XP_006712106.1 | 2 | 2204 | ||
XM_006712044.3 | XP_006712107.1 | 2 | 2204 | ||
XM_011532953.2 | XP_011531255.1 | 2 | 2204 | ||
XM_011532955.2 | XP_011531257.1 | 2 | 149 | ||
GenBank mRNA | AK001778.1 | - | 2 | 106 | |
AK095303.1 | - | 2 | 1983 | ||
AK289486.1 | - | 2 | 135 | ||
BC035315.1 | - | 2 | 134 | ||
BC064423.1 | - | 3 | 362 | 5752279 | |
BI765911.1 | - | 2 | 217 | ||
DA044670.1 | - | 3 | 384 | ||
DB024675.1 | - | 3 | 430 |