Gene Transcripts

Gene Symbol
C2orf83
Entrez Gene ID
56918
Gene Name
chromosome 2 open reading frame 83
Gene Aliases
Chromosome Location
Chr. 2: 227610090 - 227633320 on Build GRCh38
UniGene ID
Hs.283092
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162483.1 NP_001155955.1 4 757
NM_020161.3 NP_064546.3 3 662
GenBank mRNA AK055355.1 - 4 757
BC131618.1 - 3 500 40116809

View Full Product Details