Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146056.2 | NP_001139528.1 | 6 | 1568 | |
NM_001146083.2 | NP_001139555.1 | 7 | 1647 | ||
NM_024298.4 | NP_077274.3 | 8 | 1774 | ||
XM_011527299.2 | XP_011525601.1 | 8 | 1978 | ||
XM_011527300.2 | XP_011525602.1 | 8 | 1369 | ||
GenBank mRNA | AK055908.1 | - | 8 | 1707 | |
AK122789.1 | - | 9 | 1492 | ||
AK123290.1 | - | 2 | 2221 | ||
AK293199.1 | - | 6 | 1178 | ||
AK298689.1 | - | 6 | 1254 | ||
AK311493.1 | - | 7 | 1050 | ||
BC002512.2 | - | 7 | 1224 | ||
BC003164.1 | - | 8 | 1305 | 2958347 | |
BC015857.1 | - | 7 | 1211 | 4671596 | |
EU016381.1 | - | 7 | 1226 | ||
S82470.1 | - | 6 | 1080 |