Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111298.2 | NP_001104768.2 | 8 | 1419 | |
NM_001286360.1 | NP_001273289.1 | 6 | 1245 | ||
NM_173672.4 | NP_775943.1 | 7 | 1341 | ||
NR_104429.1 | 8 | 1461 | |||
XM_011535765.2 | XP_011534067.1 | 7 | 783 | ||
GenBank mRNA | AK293674.1 | - | 8 | 913 | |
AK315790.1 | - | 7 | 814 | ||
BC038716.1 | - | 7 | 800 | 5268756 | |
DQ363562.1 | - | 7 | 1341 | ||
DQ423529.1 | - | 8 | 1419 |