Gene Transcripts

Gene Symbol
SPHK1
Entrez Gene ID
8877
Gene Name
sphingosine kinase 1
Gene Aliases
SPHK
Chromosome Location
Chr. 17: 76382977 - 76387860 on Build GRCh38
UniGene ID
Hs.68061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142601.1 NP_001136073.1 5 745
NM_001142602.1 NP_001136074.1 4 790
NM_021972.3 NP_068807.2 5 787
NM_182965.2 NP_892010.2 5 1108
XM_005257766.2 XP_005257823.1 5 1100
GenBank mRNA AF200328.1 - 4 371
AF238083.1 - 5 689
AF266756.1 - 4 622
AK022402.1 - 5 745
AK023393.1 - 4 702
AK095578.1 - 4 1430
AK122781.1 - 7 1162
AK225250.1 - 5 708
AK225258.1 - 5 708
AK292294.1 - 5 832
AK292614.1 - 4 706
BC004112.2 - 1 62
BC008040.2 - 5 682 3831657
BC009419.1 - 5 775 3506924
BC014439.1 - 5 1045 4871343
BC030553.1 - 5 1067 5213270

View Full Product Details