Gene Transcripts
- Gene Symbol
- PRR22
- Entrez Gene ID
- 163154
- Gene Name
- proline rich 22
- Gene Aliases
- Chromosome Location
- Chr. 19: 5782960 - 5784765 on Build GRCh38
- UniGene ID
- Hs.631838
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC008985.1 |
- |
1 |
462 |
|
BC023278.1 |
- |
1 |
462 |
4213314 |
View Full Product Details