Gene Transcripts

Gene Symbol
AOX3P-AOX2P
Entrez Gene ID
107161151
Gene Name
AOX3P-AOX2P readthrough
Gene Aliases
AOX3/AOX3L1
Chromosome Location
Chr. 2: 200695723 - 200794915 on Build GRCh38
UniGene ID
Hs.148269
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_135011.1 30 4160
NR_135012.1 29 3653
GenBank mRNA JX431285.1 - 29 3460
JX431286.1 - 30 3967

Gene Symbol
LOC101927795
Entrez Gene ID
101927795
Gene Name
uncharacterized LOC101927795
Gene Aliases
Chromosome Location
Chr. 2: 200780495 - 200812170 on Build GRCh38
UniGene ID
Hs.567870
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110275.1 4 597
GenBank mRNA AY245869.1 - 5 585

View Full Product Details