Gene Transcripts

Gene Symbol
PADI1
Entrez Gene ID
29943
Gene Name
peptidyl arginine deiminase 1
Gene Aliases
HPAD10, PAD1, PDI, PDI1
Chromosome Location
Chr. 1: 17205123 - 17246006 on Build GRCh38
UniGene ID
Hs.412941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013358.2 NP_037490.2 15, 16 1853
XM_017001101.1 XP_016856590.1 15, 16 1756
XM_017001102.1 XP_016856591.1 15, 16 1756
GenBank mRNA AB033768.2 - 15, 16 1844
AK026652.1 - 15, 16 1855
AK127515.1 - 1, 2 1132
AK225541.1 - 15, 16 1855
AK293275.1 - 5, 6 607
AK298892.1 - 4, 5 387
AK314644.1 - 15, 16 1854
BC130574.1 - 15, 16 1832 40146605
BC136402.1 - 15, 16 1832 9020389

View Full Product Details