Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005934.3 | NP_005925.2 | 8 | 1411 | |
XM_011528021.1 | XP_011526323.1 | 8 | 1385 | ||
XM_011528022.1 | XP_011526324.1 | 8 | 1268 | ||
XM_011528023.1 | XP_011526325.1 | 7 | 1259 | ||
XM_017026819.1 | XP_016882308.1 | 8 | 1967 | ||
GenBank mRNA | D14539.1 | - | 8 | 1411 | |
L04285.1 | - | 7 | 1247 |